Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Bromopropyl)trimethoxysilane 98.0+%, TCI America™
SDP

Catalog No. B328925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B328925G 25 g
B32895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B328925G Supplier TCI America Supplier No. B328925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51826-90-5
Molecular Formula C6H15BrO3Si
Molecular Weight (g/mol) 243.172
MDL Number MFCD00053894
InChI Key GLISZRPOUBOZDL-UHFFFAOYSA-N
PubChem CID 103984
IUPAC Name 3-bromopropyl(trimethoxy)silane
SMILES CO[Si](CCCBr)(OC)OC

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 g
Formula Weight 243.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (3-Bromopropyl)trimethoxysilane
EINECSNumber (2)-2079
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.