Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromopropylamine Hydrobromide 98.0+%, TCI America™
SDP

Catalog No. p-7118958 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Catalog No. B126325G

Supplier: TCI America B126325G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5003-71-4
Molecular Formula C3H9Br2N
Molecular Weight (g/mol) 218.92
MDL Number MFCD00012912
InChI Key PQIYSSSTRHVOBW-UHFFFAOYSA-N
Synonym 3-bromopropylamine hydrobromide, 3-bromopropan-1-amine hydrobromide, 3-aminopropyl bromide hydrobromide, 1-propanamine, 3-bromo-, hydrobromide, 3-bromopropylammonium bromide, 3-bromopropanamine hydrobromide, 1-amino-3-bromopropane hydrobromide, 3-bromo-1-propanamine, hydrobromide, 3-brominepropylaminehydrobromide, 3-bromopropylamine, hydrobromide
PubChem CID 78701
IUPAC Name 3-bromopropan-1-amine;hydrobromide
SMILES C(CN)CBr.Br

Specifications

Melting Point 174°C
Color White
Quantity 25 g
Formula Weight 218.92
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromopropylamine Hydrobromide
RTECSNumber UH9925000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.