Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromoquinoline, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
5332-24-1
C9H6BrN
208.06
ZGIKWINFUGEQEO-UHFFFAOYSA-N
quinoline, 3-bromo,3-bromoquinliine,3-bromo-quinoline,3-bromo quinoline,3-bromoquinolin,pubchem5814,3-bromoquinoline,acmc-1ax3i
21413
3-bromoquinoline
C1=CC=C2C(=C1)C=C(C=N2)Br
5332-24-1
C9H6BrN
208.06
ZGIKWINFUGEQEO-UHFFFAOYSA-N
quinoline, 3-bromo,3-bromoquinliine,3-bromo-quinoline,3-bromo quinoline,3-bromoquinolin,pubchem5814,3-bromoquinoline,acmc-1ax3i
21413
3-bromoquinoline
C1=CC=C2C(=C1)C=C(C=N2)Br
Infrared Spectrum Authentic
Boiling Point 274.0°C to 276.0°C
Melting Point 13.0°C to 15.0°C
Specific Gravity 1.53
Packaging Glass bottle
Refractive Index 1.6630 to 1.6650
Density 1.5300g/mL
Color Colorless to Yellow
Flash Point >112°C
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 20, 363
Formula Weight 208.06
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Bromoquinoline
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-237-1