Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromothiophene-2-carboxaldehyde, 97%

Catalog No. AC456770010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456770010 1 g
AC456770050 5 g
AC456770250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC456770010 Supplier Thermo Scientific Chemicals Supplier No. 456770010

CAS: 930-96-1 | C5H3BrOS | 191.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-96-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00126680
InChI Key BCZHCWCOQDRYGS-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-carboxaldehyde, 3-bromo-2-formylthiophene, 3-bromo-2-thiophenecarbaldehyde, 3-bromo-2-thiophene carboxaldehyde, 3-bromo-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 3-bromo, bromothiophenecarbaldehyde, 3-bromothiophene-2-aldehyde, pubchem5327, acmc-209rjz
PubChem CID 2797079
IUPAC Name 3-bromothiophene-2-carbaldehyde
SMILES BrC1=C(SC=C1)C=O

Specifications

Melting Point 24.0°C to 25.0°C
Density 1.8100g/mL
Boiling Point 92.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.81
Formula Weight 191.05
Percent Purity 97%
Chemical Name or Material 3-Bromothiophene-2-carboxaldehyde
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.