Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromothiophene, 97%

Catalog No. aaa1402209
Click to view available options
Quantity:
10 g

CAS: 872-31-1 | C4H3BrS | 163.03 g/mol

3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, μ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, µ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-31-1
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005464
InChI Key XCMISAPCWHTVNG-UHFFFAOYSA-N
Synonym 3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
PubChem CID 13383
IUPAC Name 3-bromothiophene
SMILES BrC1=CSC=C1

Specifications

Density 1.722
Boiling Point 157°C to 158°C
Flash Point 63°C (145°F)
Odor Pungent
Refractive Index 1.591
Quantity 10 g
UN Number UN2810
Beilstein 105338
Solubility Information Immiscible with water.
Formula Weight 163.04
Percent Purity 97%
Chemical Name or Material 3-Bromothiophene
Show More Show Less
Hazard Category H226-H301-H310+H330-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H330-H227-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 212-821-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.