Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromothiophene 97.0+%, TCI America™
SDP

Catalog No. B1067250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1067250G 250 g
B106725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1067250G Supplier TCI America Supplier No. B1067250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 872-31-1
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005464
InChI Key XCMISAPCWHTVNG-UHFFFAOYSA-N
Synonym 3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
PubChem CID 13383
IUPAC Name 3-bromothiophene
SMILES BrC1=CSC=C1

Specifications

Boiling Point 158°C
Quantity 250 g
UN Number 1992
Formula Weight 163.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.