Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Buten-2-ol, 97%, Acros Organics

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
598-32-3
C4H8O
72.11
MFCD00004543
MKUWVMRNQOOSAT-UHFFFAOYSA-N
3-buten-2-ol,1-buten-3-ol,methyl vinylcarbinol,3-butene-2-ol,3-hydroxy-1-butene,propenol, 1-methyl,methyl vinyl carbinol,1-methyl-2-propenol,2-methyl-2-propenol,1-methylallyl alcohol
11716
but-3-en-2-ol
CC(C=C)O
598-32-3
C4H8O
72.11
MFCD00004543
MKUWVMRNQOOSAT-UHFFFAOYSA-N
3-buten-2-ol,1-buten-3-ol,methyl vinylcarbinol,3-butene-2-ol,3-hydroxy-1-butene,propenol, 1-methyl,methyl vinyl carbinol,1-methyl-2-propenol,2-methyl-2-propenol,1-methylallyl alcohol
11716
but-3-en-2-ol
CC(C=C)O
Infrared Spectrum Authentic
Melting Point -100.0°C
Packaging Glass bottle
Density 0.8300g/mL
Boiling Point 96.0°C to 98.0°C
Quantity 5mL
Flash Point 16°C
Color Colorless
Refractive Index 1.4130 to 1.4150
Specific Gravity 0.83
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula H2C=CHCH(OH)CH3
Solubility Information Solubility in water: soluble
Formula Weight 72.11
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Buten-2-ol
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

EINECSNumber : 209-929-8