Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Buten-2-ol, 97%

Catalog No. AC387070250
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle

CAS: 598-32-3 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-32-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00004543
InChI Key MKUWVMRNQOOSAT-UHFFFAOYNA-N
Synonym 3-buten-2-ol, 1-buten-3-ol, methyl vinylcarbinol, 3-butene-2-ol, 3-hydroxy-1-butene, propenol, 1-methyl, methyl vinyl carbinol, 1-methyl-2-propenol, 2-methyl-2-propenol, 1-methylallyl alcohol
PubChem CID 11716
IUPAC Name but-3-en-2-ol
SMILES CC(O)C=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless
Density 0.8300g/mL
Boiling Point 96.0°C to 98.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH(OH)CH3
Refractive Index 1.4130 to 1.4150
Quantity 25 mL
Specific Gravity 0.83
Solubility Information Solubility in water: soluble
Formula Weight 72.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Buten-2-ol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 209-929-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.