Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (±)-3-Butyn-2-ol, 98%

Manufacturer:  Alfa Aesar™ A1188114

Quantity 25g

 View more versions of this product

Catalog No. AAA1188114


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2028-63-9
70.09
UN2929
Methyl ethynyl carbinol; 3-Hydroxy-1-butyne
C4H6O
MFCD00004541
635722
(±)-3-Butyn-2-ol

Specifications

2028-63-9
70.09
UN2929
Methyl ethynyl carbinol; 3-Hydroxy-1-butyne
25g
(±)-3-Butyn-2-ol
C4H6O
MFCD00004541
635722
108° to 109°C
28°C (82°F)
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H310-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

RTECSNumber : ES0709800

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit