Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carbethoxy-4-piperidone hydrochloride, 98%

Catalog No. AC108330250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC108330250 25 g
AC108330050 5 g
AC108331000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC108330250 Supplier Thermo Scientific Chemicals Supplier No. 108330250
Add to Cart

CAS: 4644-61-5 | C8H13NO3·HCl | 207.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4644-61-5
Molecular Formula C8H13NO3·HCl
Molecular Weight (g/mol) 207.66
MDL Number MFCD00043267
InChI Key RTYXWLMQWSFXNI-UHFFFAOYSA-N
Synonym ethyl 4-piperidone-3-carboxylate hydrochloride, ethyl 4-oxopiperidine-3-carboxylate hydrochloride, 3-carbethoxy-4-piperidone hydrochloride, ethyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-carbethoxy-4-piperidone hcl, 3-ethoxycarbonyl-4-piperidone hydrochloride, ethyl 4-piperidone-3-carboxylate hcl, ethyl 4-piperidone-3-carboxylate, hcl, 3-piperidinecarboxylic acid, 4-oxo-, ethyl ester, hydrochloride, 4-oxo-piperidine-3-carboxylic acid ethyl ester hydrochloride
PubChem CID 107315
IUPAC Name ethyl 4-oxopiperidine-3-carboxylate;hydrochloride
SMILES CCOC(=O)C1CNCCC1=O.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 169.0°C to 175.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Formula Weight 207.66
Percent Purity 98%
Chemical Name or Material 3-Carbethoxy-4-piperidone hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-073-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.