Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carboxy-4-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193796 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 3-Carboxy-4-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 872460-12-3
Melting Point 219°C
Color White
Molecular Formula C7H6BFO4
Quantity 1 g
MDL Number MFCD03095129
Synonym 3-carboxy-4-fluorophenylboronic acid, 3-carboxy-4-fluorobenzeneboronic acid, 5-dihydroxyboranyl-2-fluorobenzoic acid, 4-fluoro-3-carboxyphenylboronic acid, benzoic acid, 5-borono-2-fluoro, 3-carboxyl-4-fluorophenylboronic acid, 5-borono-2-fluoro-benzoic acid, akos brn-0540, pubchem16787, acmc-209qj6
InChI Key DGORTXQWDDXSIQ-UHFFFAOYSA-N
SMILES OB(O)C1=CC(C(O)=O)=C(F)C=C1
IUPAC Name 5-(dihydroxyboranyl)-2-fluorobenzoic acid
Molecular Weight (g/mol) 183.93
PubChem CID 2763242
Formula Weight 183.93
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.