Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carboxy-5-nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193904
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 3-Carboxy-5-nitrophenylboronic Acid (contains varying amounts of Anhydride)
CAS 101084-81-5
Melting Point 229°C
Color White-Yellow
Molecular Formula C7H6BNO6
Quantity 1 g
MDL Number MFCD00757433
Synonym 3-carboxy-5-nitrophenylboronic acid, 3-nitro-5-carboxyphenylboronic acid, 5-nitro-3-carboxyphenylboronic acid, 3-carboxy-5-nitrophenyl boronic acid, 3-carboxy-5-nitrobenzeneboronic acid, benzoic acid, 3-borono-5-nitro, 3-dihydroxyboryl-5-nitrobenzoic acid, 3-dihydroxyboranyl-5-nitrobenzoic acid, 3-borono-5-nitro-benzoic acid, pubchem5139
InChI Key WNIFCLWDGNHGMX-UHFFFAOYSA-N
SMILES B(C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O)(O)O
IUPAC Name 3-borono-5-nitrobenzoic acid
Molecular Weight (g/mol) 210.936
PubChem CID 2773315
Formula Weight 210.94
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.