Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Carene, 90%, stabilized, ACROS Organics™

Click to view available options
Quantity:
25mL
1L
Packaging:
Glass bottle
13466-78-9,138-86-3,127-91-3,99-86-5
C10H16
136.23
MFCD00001315
BQOFWKZOCNGFEC-UHFFFAOYSA-N
3-carene,carene,3,7,7-trimethylbicyclo 4.1.0 hept-3-ene,delta-3-carene,monoterpenes,alpha-carene,bicyclo 4.1.0 hept-3-ene, 3,7,7-trimethyl,+--delta3-carene,+ car-3-ene,+--3-carene
26049
CHEBI:35661
4,7,7-trimethylbicyclo[4.1.0]hept-3-ene
CC1=CCC2C(C1)C2(C)C
13466-78-9,138-86-3,127-91-3,99-86-5
C10H16
136.23
MFCD00001315
BQOFWKZOCNGFEC-UHFFFAOYSA-N
3-carene,carene,3,7,7-trimethylbicyclo 4.1.0 hept-3-ene,delta-3-carene,monoterpenes,alpha-carene,bicyclo 4.1.0 hept-3-ene, 3,7,7-trimethyl,+--delta3-carene,+ car-3-ene,+--3-carene
26049
CHEBI:35661
4,7,7-trimethylbicyclo[4.1.0]hept-3-ene
CC1=CCC2C(C1)C2(C)C
Density 0.8640g/mL
Flash Point 45°C
Refractive Index 1.4720 to 1.474
Boiling Point 166°C to 170°C
Specific Gravity 0.864
Packaging Glass bottle
Stabilizer 0.01% alpha-Tocopherol
Quantity 25mL
Color Colorless to Yellow
Infrared Spectrum Authentic
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Beilstein 05, 94
Merck Index 15, 1835
Solubility Information Solubility in water: insoluble
Formula Weight 136.23
Physical Form Liquid
Percent Purity ≥88%
Chemical Name or Material 3-Carene
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning