Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-3-Chloro-1,2-propanediol, 98%

Catalog No. AAA161430E
Click to view available options
Quantity:
500 g
2500 g

CAS: 96-24-2 | C3H7ClO2 | 110.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-24-2
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.54
MDL Number MFCD00004712
InChI Key SSZWWUDQMAHNAQ-UHFFFAOYNA-N
Synonym 3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin
PubChem CID 7290
ChEBI CHEBI:18721
IUPAC Name 3-chloropropane-1,2-diol
SMILES OCC(O)CCl

Specifications

Melting Point -40°C
Density 1.322
Boiling Point 213°C
Flash Point 137°C (278°F)
Refractive Index 1.48
Quantity 2500 g
UN Number UN2689
Beilstein 635684
Sensitivity Hygroscopic
Merck Index 14,2145
Formula Weight 110.54
Percent Purity 98%
Chemical Name or Material (+/-)-3-Chloro-1,2-propanediol
Show More Show Less
Hazard Category H300+H330-H311-H315-H319-H335-H340-H350-H360-H372
Hazard Statement GHS H Statement
H300-H360-H351-H318-H312-H332-H315-H335
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing cancer.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: GLYCEROL ALPHA-MONOCHLOROHYDRIN
EINECSNumber 202-492-4
RTECSNumber TY4025000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.