Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-1-butene 98.0+%, TCI America™
SDP

Catalog No. C0581100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0581100G 100 g
C058125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0581100G Supplier TCI America Supplier No. C0581100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 563-52-0
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00000869
InChI Key VZGLVCFVUREVDP-UHFFFAOYSA-N
Synonym 3-chloro-1-butene, 1-butene, 3-chloro, 1-methylallyl chloride, alpha-methallyl chloride, alpha-methylallyl chloride, .alpha.-methallyl chloride, gamma-chloro-alpha-butylene, 2-chloro-3-butene, .alpha.-methylallyl chloride, .gamma.-chloro-.alpha.-butylene
PubChem CID 11242
IUPAC Name 3-chlorobut-1-ene
SMILES CC(C=C)Cl

Specifications

Color Colorless
Boiling Point 64°C
Quantity 100 g
UN Number 1993
Formula Weight 90.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-1-butene
EINECSNumber (2)-0117
RTECSNumber EM4261000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.