Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-1-propanol 98.0+%, TCI America™
SDP

Catalog No. C0270100G
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 627-30-5
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.538
MDL Number MFCD00002943
InChI Key LAMUXTNQCICZQX-UHFFFAOYSA-N
Synonym 3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1
PubChem CID 12313
IUPAC Name 3-chloropropan-1-ol
SMILES C(CO)CCl

Specifications

Color Yellow
Boiling Point 160°C
Quantity 100 g
UN Number 2849
Formula Weight 94.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-1-propanol
EINECSNumber (2)-2002
RTECSNumber UA8930000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.