Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,4,5,6-tetrafluoropyridine 98.0+%, TCI America™
SDP

Catalog No. C154925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C154925G 25 g
C15495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C154925G Supplier TCI America Supplier No. C154925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1735-84-8
Molecular Formula C5ClF4N
Molecular Weight (g/mol) 185.51
MDL Number MFCD00010624
InChI Key UXUZMYHSICIOQT-UHFFFAOYSA-N
Synonym pyridine, 3-chloro-2,4,5,6-tetrafluoro, 3-chlorotetrafluoropyridine, 2-chloro-2,4,5,6-tetrafluoropyridine, pubchem9226, 2,3,4,6-tetrafluoro-5-chloropyridine, 5-chloro-2,3,4,6-tetrafluoropyridine, 3-chloro-2,4,5,6-tetrafluoro-pyridine, pyridine,3-chloro-2,4,5,6-tetrafluoro, 3-chloro-2,4,5,6-tetrafluoropyridine, 3-chloranyl-2,4,5,6-tetrakis fluoranyl pyridine
PubChem CID 137183
IUPAC Name 3-chloro-2,4,5,6-tetrafluoropyridine
SMILES FC1=NC(F)=C(Cl)C(F)=C1F

Specifications

Color Yellow
Boiling Point 71°C
Quantity 25 g
Formula Weight 185.51
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2,4,5,6-tetrafluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.