Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2,6-difluorobenzaldehyde, 97%, Thermo Scientific™

Catalog No. AAB2355703
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190011-87-1
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.55
MDL Number MFCD01631323
InChI Key HFKZZEDGXXYRDW-UHFFFAOYSA-N
Synonym benzaldehyde, 3-chloro-2,6-difluoro, pubchem2619, timtec-bb sbb003775, ksc495e6f, acmc-1c285, benzaldehyde,3-chloro-2,6-difluoro, 3-chloro-2,6-difluorobenzaldehyde, 3-chloranyl-2,6-bis fluoranyl benzaldehyde, 3-chloro-2,6-difluoro benzaldehyde
PubChem CID 736338
IUPAC Name 3-chloro-2,6-difluorobenzaldehyde
SMILES FC1=CC=C(Cl)C(F)=C1C=O

Specifications

Melting Point 44°C to 46°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 176.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-2,6-difluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.