Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-fluoro-5-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAH6640003
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AAH6640003

Supplier: Thermo Scientific Chemicals H6640003

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209328-72-3
Molecular Formula C5H3ClFNO
Molecular Weight (g/mol) 147.533
MDL Number MFCD08061311
InChI Key IETLNKVWDZYCPN-UHFFFAOYSA-N
PubChem CID 22451555
IUPAC Name 5-chloro-6-fluoropyridin-3-ol
SMILES C1=C(C=NC(=C1Cl)F)O

Specifications

Quantity 1 g
Beilstein 13815075
Synonym 3-chloro-2-fluoro-5-hydroxypyridine, 3-pyridinol, 5-chloro-6-fluoro, 5-chloro-6-fluoro-3-pyridinol, pubchem6262, 5-chloro-6-fluoro-pyridin-3-ol, 3-pyridinol,5-chloro-6-fluoro, 3-pyridinol, 5-chloro-6-fluoro-9ci, 2-fluoro-3-chloro-5-hydroxypyridine
Formula Weight 147.53
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Chloro-2-fluoro-5-hydroxypyridine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.