Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-fluorobenzoic acid, 98%, Thermo Scientific™

Catalog No. AC399290010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 161957-55-7
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.56
InChI Key FCSSYEWURMTUSM-UHFFFAOYSA-N
Synonym 2-fluoro-3-chlorobenzoic acid, 3-chloro-2-fluoro benzoic acid, 3-chloro-2-fluoro-benzoic acid, 3-chloro-2-fluorobenzoicacid, 3-chloro-2-fluorbenzoic acid, pubchem1332, acmc-209dnj, intermediates-zcf02219, benzoic acid, chlorofluoro, ksc112c3l
PubChem CID 302931
IUPAC Name 3-chloro-2-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)F)C(=O)O

Specifications

Melting Point 177.0°C to 180.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula ClC6H3(F)COOH
Quantity 1 g
Formula Weight 174.56
Percent Purity 98%
Chemical Name or Material 3-Chloro-2-fluorobenzoic acid
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.