Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193886 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 3-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 352535-82-1
Melting Point 247°C
Color White
Molecular Formula C6H5BClFO2
Quantity 1 g
MDL Number MFCD05664224
Synonym 3-chloro-2-fluorophenyl boronic acid, 3-chloro-2-fluorobenzeneboronic acid, boronic acid, 3-chloro-2-fluorophenyl, boronic acid,b-3-chloro-2-fluorophenyl, acmc-209ifa, 3-chloro-2-fluorophenyl boranediol, 3-chloro-2-fluorophenyl boronicacid, 2-fluoro-3-chlorophenylboronic acid, 3-chloro-2-fluorophenylboronic acid
InChI Key SUYRGLRWMPEARP-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CC(Cl)=C1F
IUPAC Name (3-chloro-2-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.36
PubChem CID 16217158
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.