Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-fluorobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. C24825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24825G 5 g
C248225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24825G Supplier TCI America Supplier No. C24825G

Chemical Identifiers

CAS 192702-01-5
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.47
MDL Number MFCD01631551
InChI Key GGTQWWTYUKXFPP-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzyl bromide, 4-bromomethyl-2-chloro-1-fluorobenzene, alpha-bromo-3-chloro-4-fluorotoluene, 1-bromomethyl-3-chloro-4-fluorobenzene, 3-chloro-4-fluorobenzylbromide, 3-chloro-4-fluoro benzyl bromide, benzene, 4-bromomethyl-2-chloro-1-fluoro, pubchem2130, acmc-209ewf, 4-bromomethyl-2-chloro-1-fluoro-benzene
PubChem CID 2757542
IUPAC Name 4-(bromomethyl)-2-chloro-1-fluorobenzene
SMILES FC1=CC=C(CBr)C=C1Cl

Specifications

Color Yellow
Boiling Point 65°C
Quantity 5 g
UN Number 3265
Formula Weight 223.47
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-fluorobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.