Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-fluorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC431990500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431990500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431990500 Supplier Thermo Scientific Chemicals Supplier No. 431990500

CAS: 413589-34-1 | C6H3BrClFMg | 233.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 413589-34-1 , 109-99-9
Molecular Formula C6H3BrClFMg
Molecular Weight (g/mol) 233.75
MDL Number MFCD02260212
InChI Key XRNCDMQVKMYTAM-UHFFFAOYSA-M
Synonym bromo 3-chloro-4-fluorophenyl magnesium, 3-chloro-4-fluorophenylmagnesium bromide, xrncdmqvkmytam-uhfffaoysa-m, 3-chloro-4-fluorophenyl magnesium bromide, magnesium,1-chloro-2-fluorobenzene-5-ide,bromide, 3-chloro-4-fluorophenylmagnesium bromide, 0.50 m in 2-methf, 3-chloro-4-fluorophenylmagnesium bromide 0.5 m in tetrahydrofuran, 3-chloro-4-fluorophenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
PubChem CID 2778256
IUPAC Name magnesium(2+) 3-chloro-4-fluorobenzen-1-ide bromide
SMILES FC1=CC=C([Mg]Br)C=C1Cl

Specifications

CAS Min % 9.68, 85.48
CAS Max % 14.52, 90.32
Color Yellow to Brown
Density 0.9660g/mL
Boiling Point 65.0°C
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Quantity 50 mL
Specific Gravity 0.966
Formula Weight 233.75
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-fluorophenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.