Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-formylbenzeneboronic acid, 95%, Thermo Scientific™

Catalog No. AAH27725MD
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in suzuki cross-coupling.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072952-53-4
Molecular Formula C7H6BClO3
Molecular Weight (g/mol) 184.382
MDL Number MFCD08274473
InChI Key KDIYVLXLPNHJRZ-UHFFFAOYSA-N
Synonym 3-chloro-4-formylphenyl boronic acid, 3-chloro-4-formylbenzeneboronic acid, acmc-2098uy, ksc496a3d, 3-chloro-4-formylphenyl boronicacid, 3-chloro-4-formyl phenylboronic acid, 4-borono-2-chlorobenzaldehyde, 3-chloro-4-formyl-phenyl boronic acid
PubChem CID 42614540
IUPAC Name (3-chloro-4-formylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C=O)Cl)(O)O

Specifications

Melting Point 210°C to 214°C
Quantity 250 mg
Beilstein EM>3
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 184.39
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-formylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.