Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193848 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 3-Chloro-4-methylphenylboronic Acid (contains varying amounts of Anhydride)
Melting Point 247°C
CAS 175883-63-3
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C7H8BClO2
MDL Number MFCD04039010
Synonym 3-chloro-4-methylphenyl boronic acid, 3-chloro-4-methylbenzeneboronic acid, 3-chloro-4-methylphenylboronicacid, 3-chloro-p-tolylboronic acid, boronic acid, 3-chloro-4-methylphenyl, pubchem5132, 3-chloro-4-methyl-phenyl boronic acid, acmc-209eaq, ksc489k9f, 3-chloro-4-methylphenyl-boronic acid
InChI Key YTJUYWRCAZWVSX-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1Cl)B(O)O
IUPAC Name (3-chloro-4-methylphenyl)boronic acid
Molecular Weight (g/mol) 170.40
PubChem CID 3854610
Formula Weight 170.40
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.