Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-morpholino-1,2,5-thiadiazole 98.0+%, TCI America™
SDP

Catalog No. C218925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C218925G 25 g
C21895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C218925G Supplier TCI America Supplier No. C218925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 30165-96-9
Molecular Formula C6H8ClN3OS
Molecular Weight (g/mol) 205.66
MDL Number MFCD00274160
InChI Key LAUCCQWGVCJGFT-UHFFFAOYSA-N
PubChem CID 121645
IUPAC Name 4-(4-chloro-1,2,5-thiadiazol-3-yl)morpholine
SMILES C1COCCN1C2=NSN=C2Cl

Specifications

Melting Point 46°C
Color Yellow
Quantity 25 g
Formula Weight 205.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-morpholino-1,2,5-thiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.