Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-L-tyrosine, 97%, Thermo Scientific™

Catalog No. aal07675md
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-93-0
Molecular Formula C9H10ClNO3
Molecular Weight (g/mol) 215.63
MDL Number MFCD00045865
InChI Key ACWBBAGYTKWBCD-JLDDOWRYNA-N
Synonym 3-chlorotyrosine, 3-chloro-l-tyrosine, 3-cl-tyr-oh, unii-sy44c9mina, sy44c9mina, s-2-amino-3-3-chloro-4-hydroxyphenyl propanoic acid, 2s-2-amino-3-3-chloro-4-hydroxyphenyl propanoic acid, pubchem11980, 3-chloro-4-hydroxy-l-phenylalanine
PubChem CID 110992
ChEBI CHEBI:53678
IUPAC Name (2S)-2-amino-3-(3-chloro-4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(Cl)=C1)C(O)=O

Specifications

Melting Point ∽250°C (decomposition)
Beilstein 2941263
Quantity 250 mg
Formula Weight 215.64
Percent Purity 97%
Chemical Name or Material 3-Chloro-L-tyrosine
EINECSNumber 231-050-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.