Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloroaniline, 99%

Catalog No. p-7081850
Click to view available options
Quantity:
100 g
500 g

Catalog No. AAA1201922

Supplier: Thermo Scientific Chemicals A1201922

Only null left
Add to Cart

CAS: 108-42-9 | C6H6ClN | 127.57 g/mol

3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.

Solubility
Soluble in water. [6.8 g/L (20°C)].

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-42-9
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00007765
InChI Key PNPCRKVUWYDDST-UHFFFAOYSA-N
Synonym m-chloroaniline, 3-chlorobenzenamine, benzenamine, 3-chloro, m-chloraniline, 3-chlorophenylamine, orange gc base, m-chlorophenylamine, m-aminochlorobenzene, m-chloroaminobenzene, aniline, m-chloro
PubChem CID 7932
IUPAC Name 3-chloroaniline
SMILES NC1=CC=CC(Cl)=C1

Specifications

Melting Point -10°C
Density 1.21
Boiling Point 230°C
Quantity 100 g
Odor Characteristic
Refractive Index 1.594
UN Number UN2019
Beilstein 605969
Merck Index 14,2118
Solubility Information Soluble in water. [6.8g/L (20°C)].
Formula Weight 127.57
Percent Purity 99%
Chemical Name or Material 3-Chloroaniline
Show More Show Less
Hazard Category H301+H311-H330-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, LIQUID
EINECSNumber 203-581-0
RTECSNumber BX0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.