Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Chlorobenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
63503-60-6
C6H6BClO2
156.372
MFCD00161354
SDEAGACSNFSZCU-UHFFFAOYSA-N
3-chlorophenyl boronic acid,3-chlorobenzeneboronic acid,3-chlorophenyl boranediol,3-chlorophenylbornic acid,3-chloro benzene boronic acid,m-chlorophenylboronic acid,3-chloro-phenyl-boronic acid,boronic acid, 3-chlorophenyl,pubchem1785
2734323
(3-chlorophenyl)boronic acid
B(C1=CC(=CC=C1)Cl)(O)O
63503-60-6
C6H6BClO2
156.372
MFCD00161354
SDEAGACSNFSZCU-UHFFFAOYSA-N
3-chlorophenyl boronic acid,3-chlorobenzeneboronic acid,3-chlorophenyl boranediol,3-chlorophenylbornic acid,3-chloro benzene boronic acid,m-chlorophenylboronic acid,3-chloro-phenyl-boronic acid,boronic acid, 3-chlorophenyl,pubchem1785
2734323
(3-chlorophenyl)boronic acid
B(C1=CC(=CC=C1)Cl)(O)O
Applications
It is employed as a reactant involved in 1,4-conjugate addition reactions with ethenesulfonamides to form arylethanesulfonamides, Suzuki-Miyaura reactions with dibromotrifluoromethylbenzene, cross-coupling reactions with diazoesters3 or potassium cyanate. It is involved in the synthesis of biarylketones and phthalides and synthesis of inhibitors including PDE4 inhibitors6 among others.

Solubility
Slightly soluble in water. soluble in ether, tetrahydrofuran,, dimethyl sulfoxide, dimethyl formamide, methanol.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Melting Point 180°C to 185°C
Quantity 25g
Beilstein 2936343
Formula Weight 156.38
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning