Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzyl alcohol, 99%

Catalog No. AC108771000
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 873-76-7 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-76-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00004652
InChI Key PTHGDVCPCZKZKR-UHFFFAOYSA-N
Synonym 4-chlorobenzyl alcohol, 4-chlorophenyl methanol, p-chlorobenzyl alcohol, benzenemethanol, 4-chloro, 4-chlorobenzenemethanol, 4-chlorobenzylalcohol, benzyl alcohol, p-chloro, 4-chlorobenzylic alcohol, unii-gf8bs53b6v, ccris 5120
PubChem CID 13397
IUPAC Name (4-chlorophenyl)methanol
SMILES C1=CC(=CC=C1CO)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 69.0°C to 72.0°C
Color Beige to White
Boiling Point 234.0°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClC6H4CH2OH
Quantity 100 g
Beilstein 06, 444
Fieser 07,59
Solubility Information Solubility in water: 2.5g/L in water (20°C)
Formula Weight 142.58
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 4-Chlorobenzyl alcohol, 99%
Show More Show Less
EINECSNumber 212-852-2
RTECSNumber DN9242000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.