Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzyl alcohol, 99+%, Thermo Scientific™

Catalog No. AC108760100
Change view
Click to view available options
:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC108760100 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC108760100 Supplier Thermo Scientific Chemicals Supplier No. 108760100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 873-63-2
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C7H7ClO
Refractive Index 1.5540 to 1.5560 (20°C, 589nm)
Linear Formula ClC6H4CH2OH
Quantity 10mL
MDL Number MFCD00004632
Synonym 3-chlorobenzyl alcohol, 3-chlorophenyl methanol, m-chlorobenzyl alcohol, benzenemethanol, 3-chloro, 3-chlorobenzylalcohol, 3-chlorophenyl methan-1-ol, m-chlorobenzylalcohol, 3-chlorobenyl alcohol, pubchem3626, acmc-209qjt
InChI Key ZSRDNPVYGSFUMD-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)CO
IUPAC Name (3-chlorophenyl)methanol
Molecular Weight (g/mol) 142.582
PubChem CID 70117
Formula Weight 142.58
Percent Purity ≥99.0%
Chemical Name or Material 3-Chlorobenzyl alcohol, 99+%
Show More Show Less
EINECSNumber 212-847-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.