Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Chloroindophenol Sodium Salt [Redox Indicator], TCI America™
SDP

Catalog No. C01881G
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 41350-02-1
Molecular Formula C12H7ClNNaO2
Molecular Weight (g/mol) 255.633
MDL Number MFCD00059876
InChI Key NTMZACARFTYJPY-UHFFFAOYSA-M
Synonym Benzenone-indo-3′C-chlorophenol Sodium Salt, o-Chlorophenol-indophenol Sodium Salt, Sodium Benzenone-indo-3′C-chlorophenol
PubChem CID 44630175
IUPAC Name sodium;2-chloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
SMILES C1=CC(=O)C=CC1=NC2=CC(=C(C=C2)[O-])Cl.[Na+]

Specifications

Color Black-Green
Quantity 1 g
Formula Weight 255.63
Physical Form Crystalline Powder
Chemical Name or Material 3′-Chloroindophenol Sodium Salt [Redox Indicator]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.