Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloroisonicotinic acid, 97%, Thermo Scientific™

Catalog No. AC433750050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 88912-27-0
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
MDL Number MFCD03094685
InChI Key MYAZXWFEMDJTFE-UHFFFAOYSA-N
Synonym 3-chloroisonicotinic acid, 3-chloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-chloro, 3-chloro-4-pyridinecarboxylicacid, pubchem5087, acmc-209qxw, 3-chloroisonicotinic cid, 3-chloro-isonicotinic acid, ksc447s3l
PubChem CID 2735816
IUPAC Name 3-chloropyridine-4-carboxylic acid
SMILES C1=CN=CC(=C1C(=O)O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 227°C to 229°C
Color Black to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 157.56
Percent Purity 97%
Physical Form Solution
Chemical Name or Material 3-Chloroisonicotinic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.