Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-(Chloromethyl)-1-methyl-1H-indazole, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 180.64 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Chloromethyl)-1-methyl-1H-indazole |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement: Causes serious eye damage. Causes severe skin burns and eye damage. |
Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction