Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Chloromethyl)-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Catalog No. CC48324CB
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 84547-64-8
Molecular Formula C5H7ClN2
Molecular Weight (g/mol) 130.575
MDL Number MFCD08690275
InChI Key DHOBZYGMEYEAEM-UHFFFAOYSA-N
PubChem CID 423923
IUPAC Name 3-(chloromethyl)-1-methylpyrazole
SMILES CN1C=CC(=N1)CCl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-chloromethyl-1-methyl-1h-pyrazole, 3-chloromethyl-1-methylpyrazole, pubchem23662, 1h-pyrazole,3-chloromethyl-1-methyl, 3-chloromethyl ;-1-methyl-1h-pyrazole, 1h-pyrazole, 3-chloromethyl-1-methyl, 1h-pyrazole, 3-chloromethyl-1-methyl-, hydrochloride
Formula Weight 130.58
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-1-methyl-1H-pyrazole
Hazard Category Danger
Hazard Statement GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
If medical advice is needed, have product container or label at hand.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.