Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloromethyl-5-nitrosalicylaldehyde 97.0+%, TCI America™
SDP

Catalog No. C130525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C130525G 25 g
C13055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C130525G Supplier TCI America Supplier No. C130525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16644-30-7
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.59
MDL Number MFCD00191331
InChI Key VPZKJFJWKLYFQD-UHFFFAOYSA-N
PubChem CID 99380
IUPAC Name 3-(chloromethyl)-2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=C(CCl)C=C(C=C1C=O)[N+]([O-])=O

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 25 g
Formula Weight 215.59
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloromethyl-5-nitrosalicylaldehyde
RTECSNumber VN5500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.