Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorophenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC397491000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 36229-42-2 | C6H4BrClMg | 215.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36229-42-2, 109-99-9
Molecular Formula C6H4BrClMg
Molecular Weight (g/mol) 215.76
MDL Number MFCD00672003
InChI Key ZIYRUWPTUPFRQU-UHFFFAOYSA-M
Synonym 3-chlorophenylmagnesium bromide, 3-chlorophenyl magnesium bromide, magnesium,bromo 3-chlorophenyl, m-chlorophenylmagnesium bromide, m-chlorophenyl magnesium bromide, 3-chloro phenyl magnesium bromide, 3-chloro-phenyl magnesium bromide, ziyruwptupfrqu-uhfffaoysa-m, grignard reagent
PubChem CID 12252967
IUPAC Name magnesium;chlorobenzene;bromide
SMILES ClC1=CC([Mg]Br)=CC=C1

Specifications

CAS Min % 11.24, 86.52
CAS Max % 13.48, 88.76
Quantity 100 mL
Linear Formula ClC6H4MgBr
Flash Point <−17°C
Solubility Information Solubility in water: reacts
Formula Weight 215.76
Concentration or Composition (by Analyte or Components) 0.5M (Argentometry), exact strength on the certificate of analysis
Identification Identity confirmed
Packaging AcroSeal™ Glass Bottle
Appearance Clear yellow to brown solution
Specific Gravity 0.96
Density 0.9600g/mL
Chemical Name or Material 3-Chlorophenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.