Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropionaldehyde Diethyl Acetal (stabilized with CaCO3) 97.0+%, TCI America™
SDP

Catalog No. C126825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C126825G 25 g
C12685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C126825G Supplier TCI America Supplier No. C126825G

Chemical Identifiers

CAS 35573-93-4
Molecular Formula C7H15ClO2
Molecular Weight (g/mol) 166.645
MDL Number MFCD00000994
InChI Key NXHONHDWVLPPCS-UHFFFAOYSA-N
Synonym 3-chloropropionaldehyde diethyl acetal, propane, 3-chloro-1,1-diethoxy, 3-chloropropionaldehyde diethylacetal, 1-chloro-3,3-diethoxypropane, .beta.-chloropropionaldehyde diethyl acetal, 3,3-diethoxypropylchloride, 3,3-diethoxypropyl chloride, 3-ethoxy-4-oxahexyl chloride, 3-chloro-1,1-diethoxy-propane, 3-chloropropanal diethyl acetal
PubChem CID 96217
IUPAC Name 3-chloro-1,1-diethoxypropane
SMILES CCOC(CCCl)OCC

Specifications

Color Yellow
Boiling Point 74°C
Quantity 25 g
UN Number 1993
Formula Weight 166.65
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropropionaldehyde Diethyl Acetal (stabilized with CaCO3)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.