Learn More
3-Chloropropyl acetate, 97+%
Description
3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.
Solubility
Soluble in water (partly).
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.12 |
| Boiling Point | 70°C to 72°C (15 mmHg) |
| Flash Point | 68°C (154°F) |
| Refractive Index | 1.429 |
| Quantity | 500 g |
| Solubility Information | Soluble in water (partly). |
| Formula Weight | 136.58 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Chloropropyl acetate |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| RTECSNumber | UA8944000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.