Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropyl acetate, 97+%

Catalog No. p-7027939
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2501314 25 g
AAB2501322 100 g
AAB2501336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2501314 Supplier Thermo Scientific Chemicals Supplier No. B2501314
Only null left
Add to Cart

CAS: 628-09-1 | C5H9ClO2 | 136.575 g/mol

3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

Solubility
Soluble in water (partly).

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-09-1
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00001000
InChI Key KPOHQIPNNIMWRL-UHFFFAOYSA-N
Synonym 1-propanol, 3-chloro-, acetate, 3-chloropropanol acetate, 3-chlorprop-1-ylacetat, gamma-chloropropyl acetate, 3-chlorprop-1-ylacetat german, acetic acid 3-chloro-propyl ester, 1-acetoxy-3-chloropropane, .gamma.-chloropropyl acetate, 3-chloro-propyl acetate, acetic acid 3-chloropropyl ester
PubChem CID 12334
IUPAC Name 3-chloropropyl acetate
SMILES CC(=O)OCCCCl

Specifications

Density 1.12
Boiling Point 70°C to 72°C (15 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.429
Quantity 25 g
Solubility Information Soluble in water (partly).
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material 3-Chloropropyl acetate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
RTECSNumber UA8944000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.