Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropylamine Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. C1062100G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 6276-54-6
Molecular Formula C3H9Cl2N
Molecular Weight (g/mol) 130.012
MDL Number MFCD00012913
InChI Key IHPRVZKJZGXTBQ-UHFFFAOYSA-N
Synonym 3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b
PubChem CID 11469095
IUPAC Name 3-chloropropan-1-amine;hydrochloride
SMILES C(CN)CCl.Cl

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 100 g
Formula Weight 130.01
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloropropylamine Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.