Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropyldichloromethylsilane 97.0+%, TCI America™
SDP

Catalog No. D199510ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D199510ML 10 mL
D19955ML 5 mL
D199525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D199510ML Supplier TCI America Supplier No. D199510ML

Chemical Identifiers

CAS 7787-93-1
Molecular Formula C4H9Cl3Si
Molecular Weight (g/mol) 191.551
MDL Number MFCD00000996
InChI Key UCJHMXXKIKBHQP-UHFFFAOYSA-N
Synonym 3-(Dichloromethylsilyl)propyl Chloride
PubChem CID 82235
IUPAC Name dichloro-(3-chloropropyl)-methylsilane
SMILES C[Si](CCCCl)(Cl)Cl

Specifications

Boiling Point 185°C
Quantity 10 mL
UN Number 2986
Formula Weight 191.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropropyldichloromethylsilane
EINECSNumber (2)-2047
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.