Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorotoluene, 97%

Catalog No. AC160450025
Click to view available options
Quantity:
100 mL
250 mL
500 mL
2.5 L
Packaging:
Glass bottle

CAS: 108-41-8 | C7H7Cl | 126.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-41-8
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000595
InChI Key OSOUNOBYRMOXQQ-UHFFFAOYSA-N
Synonym 3-chlorotoluene, m-chlorotoluene, benzene, 1-chloro-3-methyl, m-tolyl chloride, 3-chloro-1-methylbenzene, 1-methyl-3-chlorobenzene, toluene, m-chloro, meta-chlorotoluene, unii-5k8172k9tt, chlorotoluene 3-
PubChem CID 7931
IUPAC Name 1-chloro-3-methylbenzene
SMILES CC1=CC=CC(Cl)=C1

Specifications

Melting Point -48°C
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Density 1.0700g/mL
Boiling Point 160°C to 162°C
Quantity 2.5 L
Flash Point 50°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5210 to 1.5230 (20°C, 589nm)
Assay Percent Range 97%
Specific Gravity 1.07
Linear Formula CH3C6H4Cl
Beilstein 05, 291
Merck Index 15, 2175
Solubility Information Solubility in water: slightly soluble
Formula Weight 126.59
Percent Purity 97%
Chemical Name or Material 3-Chlorotoluene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 203-580-5
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.