Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyano-4-methylpyridine, 97%

Catalog No. AAL1976906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1976906 5 g
AAL1976903 1 g
AAL1976914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1976906 Supplier Thermo Scientific Chemicals Supplier No. L1976906
Add to Cart

CAS: 5444-01-9 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5444-01-9
MDL Number MFCD00234272

Specifications

Melting Point 41°C to 44°C
Boiling Point 108°C to 111°C (20 mmHg)
Odor Characteristic
Molecular Formula C7H6N2
Quantity 5 g
Beilstein 112016
Synonym 3-cyano-4-methylpyridine, 4-methylnicotinonitrile, 3-cyano-4-picoline, 4-methyl-nicotinonitrile, 3-pyridinecarbonitrile, 4-methyl, 4-methyl-3-pyridinecarbonitrile, 3-cyano-4-methylpyridine 3-cyano-4-picoline, pubchem6623, pubchem18426, 4-methyl nicotinonitrile
InChI Key XLAPHZHNODDMDD-UHFFFAOYSA-N
SMILES CC1=C(C=NC=C1)C#N
IUPAC Name 4-methylpyridine-3-carbonitrile
Molecular Weight (g/mol) 118.14
PubChem CID 227806
Formula Weight 118.14
Percent Purity 97%
Chemical Name or Material 3-Cyano-4-methylpyridine
Show More Show Less
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.