Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanothiophene 98.0+%, TCI America™
SDP

Catalog No. C134425G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1641-09-4
Molecular Formula C5H3NS
Molecular Weight (g/mol) 109.146
MDL Number MFCD00151852
InChI Key GSXCEVHRIVLFJV-UHFFFAOYSA-N
Synonym 3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h
PubChem CID 74231
IUPAC Name thiophene-3-carbonitrile
SMILES C1=CSC=C1C#N

Specifications

Color Yellow
Boiling Point 205°C
Quantity 25 g
Formula Weight 109.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Cyanothiophene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.