Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclohexyl-L-alanine, 95%, Thermo Scientific Chemicals

Catalog No. AAH6591306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6591306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6591306 Supplier Thermo Scientific Chemicals Supplier No. H6591306

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27527-05-5
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00153484
InChI Key ORQXBVXKBGUSBA-QMMMGPOBSA-N
Synonym s-2-amino-3-cyclohexylpropanoic acid, l-cyclohexylalanine, h-cha-oh, 2s-2-amino-3-cyclohexylpropanoic acid, 3-cyclohexyl-l-alanine, l-3-cyclohexylalanine, beta-cyclohexyl-alanine, s-2-amino-3-cyclohexylpropionic acid, l-beta-cyclohexylalanine, 3-cyclohexylalanine
PubChem CID 712421
IUPAC Name (2S)-2-amino-3-cyclohexylpropanoic acid
SMILES C1CCC(CC1)CC(C(=O)O)N

Specifications

Quantity 5 g
Formula Weight 171.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Cyclohexyl-L-alanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.