Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Deoxy-D-glucosone, 95%, Thermo Scientific Chemicals

Catalog No. p-7043208
Click to view available options
Quantity:
5 mg

Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents. Also used for cross-linking studies as it readily reacts with protein amino groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Frequently used as a reference for the analysis and detection of glucose degradation products and glycating agents
  • Also used for cross-linking studies as it readily reacts with protein amino groups
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4084-27-9
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD01723101
InChI Key ZGCHLOWZNKRZSN-NTSWFWBYSA-N
Synonym 3-deoxyglucosone, 3-deoxy-d-glucosone, 3-deoxy-d-erythro-hexos-2-ulose, d-3-deoxyglucosone, 3-deoxy-d-erythro-hexosulose, 3-deoxy-d-fructose, ccris 4272, 3-deoxy-2-ketoglucose, d-erythro-hexos-2-ulose, 3-deoxy, unii-exv5374vey
PubChem CID 114839
ChEBI CHEBI:60777
IUPAC Name (4S,5R)-4,5,6-trihydroxy-2-oxohexanal
SMILES C(C(C(CO)O)O)C(=O)C=O

Specifications

Color Yellow
Quantity 5 mg
Solubility Information Soluble in water,methanol or hot ethanol.
Formula Weight 162.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Deoxy-D-glucosone
RTECSNumber MQ3390000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.