Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Diethylamino-1-propanol, 95%

Catalog No. AC165410500
Click to view available options
Quantity:
5 g
25 mL
50 g
100 mL

CAS: 622-93-5 | C7H17NO | 131.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-93-5
Molecular Formula C7H17NO
Molecular Weight (g/mol) 131.22
MDL Number MFCD00002947
InChI Key WKCYFSZDBICRKL-UHFFFAOYSA-N
Synonym 3-diethylamino-1-propanol, 3-diethylamino propan-1-ol, 1-propanol, 3-diethylamino, 3-diethylamino propanol, 3-diethylamino propyl alcohol, n,n-diethyl-3-hydroxypropylamine, diethylpropanolamine, unii-u57yow5553, gamma-diaethylaminopropanol german
PubChem CID 12159
IUPAC Name 3-(diethylamino)propan-1-ol
SMILES CCN(CC)CCCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 81°C to 83°C (15.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4425 to 1.4445
Linear Formula (C2H5)2N(CH2)3OH
Quantity 50 g
Beilstein 04,288
Specific Gravity 0.88
Formula Weight 131.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Diethylamino-1-propanol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
EINECSNumber 210-759-1
RTECSNumber UB1480000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.