Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Diethylamino)propionic Acid Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D171325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D171325G 25 g
D17135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D171325G Supplier TCI America Supplier No. D171325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15674-67-6
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00012611
InChI Key QQVHFNAGPZTCBE-UHFFFAOYSA-N
Synonym 3-diethylamino propanoic acid hydrochloride, n,n-diethyl-beta-alanine hydrochloride, 3-diethylamino propionic acid hydrochloride, 3-diethylaminopropionic acid hydrochloride, 3-diethylamino-propionic acid hydrochloride, acmc-1busg, 3-diethylamino propanoicacidhydrochloride, 3-diethylamino propionicacidhydrochloride, 3-diethylamino propionic acid hcl
PubChem CID 3084241
IUPAC Name 3-(diethylamino)propanoic acid;hydrochloride
SMILES CCN(CC)CCC(=O)O.Cl

Specifications

Melting Point 146°C
Color White
Quantity 25 g
Formula Weight 181.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Diethylamino)propionic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.