Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[3-(Diethylamino)propyl]trimethoxysilane 97.0+%, TCI America™
SDP

Catalog No. D436225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D436225ML 25 mL
D43625ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D436225ML Supplier TCI America Supplier No. D436225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41051-80-3
Molecular Formula C10H25NO3Si
Molecular Weight (g/mol) 235.40
MDL Number MFCD00039882
InChI Key ZLDHYRXZZNDOKU-UHFFFAOYSA-N
Synonym N,N-Diethyl-3-(trimethoxysilyl)propylamine
PubChem CID 170458
IUPAC Name diethyl[3-(trimethoxysilyl)propyl]amine
SMILES CCN(CC)CCC[Si](OC)(OC)OC

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
Formula Weight 235.40
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material [3-(Diethylamino)propyl]trimethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.